Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
1 x 8 Stripwell
™
96 well plates
Products
Product Category
plates (12)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Plate Material
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
CLS2592
Corning
®
1 x 8 Stripwell
™
96 well plates,
high binding surface, clear
Expand
View Pricing
CLS2593
Corning
®
1 x 8 Stripwell
™
96 well plates,
medium binding surface, clear
Expand
View Pricing
CLS2485
Corning
®
1 x 8 Stripwell
™
96 well plates,
low volume, high binding surface, bulk packed, white
Expand
View Pricing
CLS2580
Corning
®
1 x 8 Stripwell
™
96 well plates,
high binding surface, without stripwell holder
Expand
View Pricing
CLS42592
Corning
®
1 x 8 Stripwell
™
96 well plates,
high binding surface, flat bottom clear (4 cavity), non-sterile
Expand
View Pricing
CLS2483
Corning
®
1 x 8 Stripwell
™
96 well plates,
low volume, high binding surface, black
Expand
View Pricing
CLS3924
Corning
®
1 x 8 Stripwell
™
96 well plates,
high binding surface w/o lid, black
Expand
View Pricing
CLS3923
Corning
®
1 x 8 Stripwell
™
96 well plates,
high bind surface, white
Expand
View Pricing
CLS2482
Corning
®
1 x 8 Stripwell
™
96 well plates,
low volume, medium binding surface, black
Expand
View Pricing
CLS43923
Corning
®
1 x 8 Stripwell
™
96 well plates,
high binding surface, flat bottom white (4 cavity), non-sterile
Expand
View Pricing
CLS2481
Corning
®
1 x 8 Stripwell
™
96 well plates,
low volume, high binding surface, clear
Expand
View Pricing
CLS2484
Corning
®
1 x 8 Stripwell
™
96 well plates,
low volume, medium binding surface, bulk packed, white
Expand
View Pricing