Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
15 mL centrifuge tubes
Products
Product Category
tubes (7)
Color
Manufacturer
Agency
Capacity (mL)
Diameter (mm)
OD (mm)
Cap
Color
Package Size
Special Grade
Natural (7)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
CLS430791
Corning
®
15 mL centrifuge tubes,
15 mL centrifuge tubes, polypropylene, conical bottom w/ CentriStar cap, sterile, natural, 500/cs
Expand
View Pricing
CLS430790
Corning
®
15 mL centrifuge tubes,
15 mL centrifuge tubes, polypropylene, conical bottom w/ CentriStar cap, rack packed, sterile, natural, 50/rack, 500/cs
Expand
View Pricing
CLS430052
Corning
®
15 mL centrifuge tubes,
conical bottom natural polypropylene, sterile, self-standing: no (rack packed), cap (plug seal), case of 500
Expand
View Pricing
CLS430055
Corning
®
15 mL centrifuge tubes,
15 mL centrifuge tubes, PET, conical bottom w/ plug seal cap, rack packed, sterile, natural, 50/rack, 500/cs
Expand
View Pricing
CLS430766
Corning
®
15 mL centrifuge tubes,
15 mL centrifuge tubes, polypropylene, conical bottom w/ plug seal cap, sterile, natural, 500/cs
Expand
View Pricing
CLS431470
Corning
®
15 mL centrifuge tubes,
15 mL centrifuge tubes, polypropylene, conical bottom w/ no cap, sterile, natural, 500/cs
Expand
View Pricing
CLS430053
Corning
®
15 mL centrifuge tubes,
conical bottom natural PET, sterile, bag of 50 (bulk packed), cap (plug seal), case of 500
Expand
View Pricing