Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper/silver
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF36840720
Copper/silver,
rod, Cu 95%/Ag 5%, 15 mm diameter, length 50 mm, temper as drawn
Expand
View Pricing
GF37355362
Copper/silver,
rod, Cu 70%/Ag 30%, 15 mm diameter, length 200 mm, temper as drawn
Expand
View Pricing
GF68236836
Copper/silver,
rod, Cu90%/Ag10%, 15 mm diameter, temper as drawn, length 50 mm
Expand
View Pricing
GF93903622
Copper/silver,
rod, Cu70%/Ag30%, 15 mm diameter, length 50 mm, temper as drawn
Expand
View Pricing
GF95852942
Copper/silver,
rod, 15 mm diameter, Cu70%/Ag30%, length 100 mm, temper as drawn
Expand
View Pricing