Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper-nickel alloy, Cu70Ni30
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
GF57925053
Copper-nickel alloy, Cu70Ni30,
200mm rod, 12.7mm diameter, as drawn
Expand
View Pricing
GF30062494
Copper-nickel alloy, Cu70Ni30,
200mm tube, 0.4mm outside diameter, 0.15mm wall thickness, 0.1mm inside diameter, hard
Expand
View Pricing
GF98499818
Copper-nickel alloy, Cu70Ni30,
foil, 300 x 300mm, 1.6mm thickness
Expand
View Pricing
GF32210749
Copper-nickel alloy, Cu70Ni30,
1000mm tube, 1.5mm outside diameter, 0.18mm wall thickness, 1.14mm inside diameter
Expand
View Pricing
GF47901157
Copper-nickel alloy, Cu70Ni30,
500mm tube, 0.4mm outside diameter, 0.15mm wall thickness, 0.1mm inside diameter, hard
Expand
View Pricing
GF25871160
Copper-nickel alloy, Cu70Ni30,
foil, 150 x 150mm, 1.6mm thickness
Expand
View Pricing
GF17444426
Copper-nickel alloy, Cu70Ni30,
500mm tube, 1.5mm outside diameter, 0.18mm wall thickness, 1.14mm inside diameter
Expand
View Pricing
GF20585606
Copper-nickel alloy, Cu70Ni30,
1000mm tube, 3.0mm outside diameter, 0.175mm wall thickness, 2.65mm inside diameter, hard
Expand
View Pricing
GF06245355
Copper-nickel alloy, Cu70Ni30,
100mm tube, 0.4mm outside diameter, 0.15mm wall thickness, 0.1mm inside diameter, hard
Expand
View Pricing
GF85089774
Copper-nickel alloy, Cu70Ni30,
100mm tube, 1.5mm outside diameter, 0.18mm wall thickness, 1.14mm inside diameter
Expand
View Pricing
GF41254950
Copper-nickel alloy, Cu70Ni30,
100mm tube, 3.0mm outside diameter, 0.175mm wall thickness, 2.65mm inside diameter, hard
Expand
View Pricing
GF09914272
Copper-nickel alloy, Cu70Ni30,
500mm tube, 3.0mm outside diameter, 0.175mm wall thickness, 2.65mm inside diameter, hard
Expand
View Pricing
GF86761547
Copper-nickel alloy, Cu70Ni30,
200mm tube, 1.5mm outside diameter, 0.18mm wall thickness, 1.14mm inside diameter
Expand
View Pricing