Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper(I) iodide
Synonym(s):
Cuprous iodide, Copper(I) iodide
Empirical Formula (Hill Notation):
CuI
CAS Number:
7681-65-4
Molecular Weight:
190.45
EC Number:
231-674-6
Products
Product Category
catalysts (1)
salts (1)
Brand
Sigma-Aldrich (5)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Reaction Type
Reagent Type
Core
Special Grade
Anhydrous (1)
Purum (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
205540
Copper(I) iodide,
98%
Expand
View Pricing
215554
Copper(I) iodide,
99.999% trace metals basis
Expand
View Pricing
792063
Copper(I) iodide,
anhydrous, 99.995% trace metals basis
Expand
View Pricing
8.18311
Copper(I) iodide,
for synthesis
Expand
View Pricing
03140
Copper(I) iodide,
purum, ≥99.5%
Expand
View Pricing