Conjugated Polymer Nanoparticles incorporating iron oxide
Products
Brand
Physical Form
Quality Segment
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Conjugated Polymer Nanoparticles incorporating iron oxide, fluorescence λem 475 nm, 100 μg/mL in H2O
Conjugated Polymer Nanoparticles incorporating iron oxide, fluorescence λem 680 nm, 100 μg/mL in H2O
Conjugated Polymer Nanoparticles incorporating iron oxide, fluorescence λem 550 nm, 100 μg/mL in H2O