<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
AD
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Conductivity Standard
Products
Brand
Supelco (14)
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (15)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
06949
Conductivity Standard,
conductivity 1000 μS/cm at 25 °C
Expand
View Pricing
06999
Conductivity Standard,
conductivity 500 μS/cm at 25 °C, ≤2.5% KCl (T)
Expand
View Pricing
06811
Conductivity Standard,
conductivity 200 μS/cm at 25 °C
Expand
View Pricing
67233
Conductivity Standard,
conductivity 10000 μS/cm at 25 °C
Expand
View Pricing
56255
Conductivity Standard,
conductivity 50000 μS/cm at 25 °C
Expand
View Pricing
59962
Conductivity Standard,
conductivity 5000 μS/cm at 25 °C
Expand
View Pricing
59777
Conductivity Standard,
conductivity 100000 μS/cm at 25 °C
Expand
View Pricing
03729
Conductivity Standard,
conductivity 200000 μS/cm at 25 °C
Expand
View Pricing
56890
Conductivity Standard,
conductivity 300 μS/cm at 25 °C
Expand
View Pricing
66337
Conductivity Standard,
conductivity 300000 μS/cm at 25 °C
Expand
View Pricing
51795
Conductivity Standard,
conductivity 2000 μS/cm at 25 °C
Expand
View Pricing
44348
Conductivity Standard,
conductivity 3000 μS/cm at 25 °C
Expand
View Pricing
91916
Conductivity Standard,
conductivity 30000 μS/cm at 25 °C
Expand
View Pricing
41303
Conductivity Standard,
conductivity 400 μS/cm at 25 °C
Expand
View Pricing
51715
Conductivity Standard,
conductivity 20000 μS/cm at 25 °C
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Français - FR
Learn More