Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Compact 4 Drawer Locking Cart
Products
Manufacturer
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
TRT51146
Compact 4 Drawer Locking Cart,
black (drawers)
Expand
View Pricing
TRT51032
Compact 4 Drawer Locking Cart,
white (drawers)
Expand
View Pricing
TRT51088
Compact 4 Drawer Locking Cart,
silver (drawers)
Expand
View Pricing
TRT51080
Compact 4 Drawer Locking Cart,
blue (drawers)
Expand
View Pricing
TRT51087
Compact 4 Drawer Locking Cart,
beige (drawers)
Expand
View Pricing