Skip to Content
Merck
vibrant-m

Codeine phosphate

vibrant-m
Synonym(s):
Codeine phosphate hemihydrate
Empirical Formula (Hill Notation):
C18H21N2O3 · H3PO4 · 0.5H2O
CAS Number:
Molecular Weight:
420.37

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade