Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cochranes atom centers for Minit molecular models
Products
Color
Manufacturer
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
Z127272
Cochranes atom centers for Minit molecular models,
nitrogen, tetrahedral, blue
Expand
View Pricing
Z127264
Cochranes atom centers for Minit molecular models,
carbon, tetrahedral, black
Expand
View Pricing
Z159255
Cochranes atom centers for Minit molecular models,
oxygen, tetrahedral, red
Expand
View Pricing
Z159948
Cochranes atom centers for Minit molecular models,
sulfur, tetrahedral, yellow
Expand
View Pricing
Z184616
Cochranes atom centers for Minit molecular models,
halogen, tetrahedral, green
Expand
View Pricing
Z185523
Cochranes atom centers for Minit molecular models,
Metal (octahedral)
Expand
View Pricing
Z127280
Cochranes atom centers for Minit molecular models,
phosphorus, tetrahedral, purple
Expand
View Pricing
Z127256
Cochranes atom centers for Minit molecular models,
carbon, 6 ring planar, black
Expand
View Pricing
Z159476
Cochranes atom centers for Minit molecular models,
hydrogen, hydrogen bond, white
Expand
View Pricing
Z129585
Cochranes atom centers for Minit molecular models,
nitrogen, 6 ring planar/azo, blue
Expand
View Pricing
Z151017
Cochranes atom centers for Minit molecular models,
nitrogen, octahedral, blue
Expand
View Pricing
Z127221
Cochranes atom centers for Minit molecular models,
oxygen, diunivalent/hydrogen bonding, red
Expand
View Pricing
Z159913
Cochranes atom centers for Minit molecular models,
oxygen, trigonal, red
Expand
View Pricing
Z159972
Cochranes atom centers for Minit molecular models,
metal, tetrahedral, silver
Expand
View Pricing