Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GF67484346
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2,
foil, 300 x 300mm, 0.6mm thickness
Expand
View Pricing
GF16601341
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2,
foil, 50 x 50mm, 0.25mm thickness
Expand
View Pricing
GF21131941
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2,
foil, 150 x 150mm, 0.25mm thickness
Expand
View Pricing
GF04679499
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2,
foil, 100 x 100mm, 0.25mm thickness
Expand
View Pricing
GF18288692
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2,
foil, 150 x 150mm, 0.6mm thickness
Expand
View Pricing
GF41732108
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2,
foil, 0.5m coil, 0.25mm thickness
Expand
View Pricing
GF88828004
Cobalt-chromium-tungsten-nickel-iron-manganese alloy, Co50Cr20W15Ni10Fe3Mn2,
foil, 300 x 300mm, 0.25mm thickness
Expand
View Pricing