Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Coated GC Packing
EC Number:
215-609-9
Products
Brand
Supelco (12)
Column Type
Quality Segment
Package Size
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
11815
Coated GC Packing,
phase SP
®
-1000, 1%, matrix 60/80 Carbopack B support, bottle of 15 g
Expand
View Pricing
11766
Coated GC Packing,
phase 5% Carbowax 20M, matrix 60/80 Carbopack B support, bottle of 15 g
Expand
View Pricing
11812-U
Coated GC Packing,
phase 5% Carbowax 20M, matrix 80/120 Carbopack B AW support, bottle of 15 g
Expand
View Pricing
11820
Coated GC Packing,
phase SP
®
-1000, 0.1%, matrix 80/100 Carbopack C support, bottle of 15 g
Expand
View Pricing
11993
Coated GC Packing,
phase 5% Carbowax 20M, matrix 40/60 Chromosorb T support, bottle of 50 g
Expand
View Pricing
11827
Coated GC Packing,
phase 0.2% Carbowax 1500, matrix 80/100 Carbopack C support, bottle of 15 g
Expand
View Pricing
11887
Coated GC Packing,
phase 4% Carbowax 20M + 0.8% KOH, matrix 60/80 Carbopack B support, bottle of 15 g
Expand
View Pricing
11814
Coated GC Packing,
phase 6.6% Carbowax 20M, matrix 80/120 Carbopack B AW support, bottle of 15 g
Expand
View Pricing
11813-U
Coated GC Packing,
phase 3% SP
®
-1500, matrix 80/120 Carbopack B support, bottle of 15 g
Expand
View Pricing
11826
Coated GC Packing,
phase 0.2% Carbowax 1500, matrix 60/80 Carbopack C support, bottle of 15 g
Expand
View Pricing
11809
Coated GC Packing,
phase 1% SP
®
-1510, matrix 60/80 Carbopack B support, bottle of 15 g
Expand
View Pricing
1505-U
Coated GC Packing,
phase 1.5% OV-225 + 1% H
3
PO
4
, matrix 60/80 Carbopack B support, bottle of 10 g
Expand
View Pricing