Skip to Content
Merck
cis-Diammineplatinum(II) dichloride crystalline

cis-Diammineplatinum(II) dichloride

cis-Diammineplatinum(II) dichloride crystalline
Synonym(s):
cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, Cisplatin
Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
EC Number:

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Activity Spectrum
Markush Class
Markush Group
Mode of Action
Showing 1-1 of 1