Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
cis
-8,11,14-Eicosatrienoic acid
Synonym(s):
Homo-γ-linolenic acid
Linear Formula:
CH
3
(CH
2
)
3
(CH
2
CH=CH)
3
(CH
2
)
6
CO
2
H
CAS Number:
1783-84-2
Molecular Weight:
306.48
EC Number:
217-233-0
Products
Product Category
lipids (2)
fatty acids (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Formula Weight
Physical Form
Purity
Format
Tissue
Quality Segment
Functional Group
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
E4504
cis
-8,11,14-Eicosatrienoic acid,
≥99%
Expand
View Pricing
69154
cis
-8,11,14-Eicosatrienoic acid,
analytical standard
Expand
View Pricing