
Cilostazol Related Compound A

Synonym(s):
6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 6-Hydroxy-1,2,3,4-tetrahydro-2-quinolinone, 6-Hydroxy-3,4-dihydrocarbstyril
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2