Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chlorpyrifos-methyl
Empirical Formula (Hill Notation):
C
7
H
7
Cl
3
NO
3
PS
CAS Number:
5598-13-0
Molecular Weight:
322.53
EC Number:
227-011-5
Products
Product Category
pesticides (1)
Brand
Supelco (1)
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
45396
Chlorpyrifos-methyl,
PESTANAL
®
, analytical standard
Expand
View Pricing