Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chloroform-d
Synonym(s):
Deuterochloroform
Linear Formula:
CDCl
3
CAS Number:
865-49-6
Molecular Weight:
120.38
EC Number:
212-742-4
Products
Product Category
solvents (11)
stable isotopes (9)
Brand
Sigma-Aldrich (11)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
151823
Chloroform-d,
99.8 atom % D
Expand
View Pricing
225789
Chloroform-d,
99.8 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing
612200
Chloroform-d,
99.8 atom % D, contains 0.05 % (v/v) TMS
Expand
View Pricing
151831
Chloroform-d,
99.8 atom % D, contains 1 % (v/v) TMS
Expand
View Pricing
434876
Chloroform-d,
99.8 atom % D, contains 0.1 % (v/v) TMS
Expand
View Pricing
530735
Chloroform-d,
≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Expand
View Pricing
416754
Chloroform-d,
≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Expand
View Pricing
151858
Chloroform-d,
"100%", 99.95 atom % D
Expand
View Pricing
570699
Chloroform-d,
≥99.8 atom % D, anhydrous
Expand
View Pricing
494275
Chloroform-d,
"100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing
431915
Chloroform-d,
"100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Expand
View Pricing