
Chloramine-T hydrate

Synonym(s):
N-Chloro-p-toluenesulfonamide sodium salt hydrate
Linear Formula:
CH3C6H4SO2N(Cl)Na · xH2O
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1