Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Certified glass inserts for 12 x 32 mm, large opening vials
Products
Product Category
inserts (4)
Height (mm)
OD (mm)
Volume (ml)
Color
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
29435-U
Certified glass inserts for 12 x 32 mm, large opening vials,
volume 0.2 mL, clear glass insert (with plastic bottom spring), O.D. × H 6 mm × 29 mm, pkg of 100 ea
Expand
View Pricing
29436-U
Certified glass inserts for 12 x 32 mm, large opening vials,
volume 0.25 mL, clear glass insert (no spring required), conical point, O.D. × H 6 mm × 31 mm, pkg of 100 ea
Expand
View Pricing
29441-U
Certified glass inserts for 12 x 32 mm, large opening vials,
volume 0.35 mL, clear glass insert (flat bottom), O.D. × H 6 mm × 31 mm, pkg of 100 ea
Expand
View Pricing
29437-U
Certified glass inserts for 12 x 32 mm, large opening vials,
volume 0.3 mL, clear glass insert, conical point, O.D. × H 6 mm × 31 mm, pkg of 100 ea
Expand
View Pricing