Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cellulose Chromatography Resin
Synonym(s):
Cellulose powder, Cotton linters
CAS Number:
9004-34-6
EC Number:
232-674-9
Products
Product Category
resins (3)
Brand
Supelco (1)
Biological Source
Physical Form
Separation Technique
Length (mm)
Quality Segment
Particle Size (µm)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
22183
Cellulose Chromatography Resin,
powder, 140-1400 mesh
Expand
View Pricing
22184
Cellulose Chromatography Resin,
acid washed, powder, 60-700 mesh
Expand
View Pricing
22182
Cellulose Chromatography Resin,
acid washed, from spruce, 100-700 mesh
Expand
View Pricing