Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cellulose acetate butyrate
CAS Number:
9004-36-8
Products
Brand
Sigma-Aldrich (5)
Manufacturer
Physical Form
Format
Quality Segment
Application
Polymer Composition
Polymer Type
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
419044
Cellulose acetate butyrate,
average M
n
~70,000
Expand
View Pricing
180963
Cellulose acetate butyrate,
Expand
View Pricing
419052
Cellulose acetate butyrate,
average M
n
~30,000
Expand
View Pricing
419036
Cellulose acetate butyrate,
average M
n
~12,000
Expand
View Pricing
419060
Cellulose acetate butyrate,
average M
n
~30,000
Expand
View Pricing
Y0001585
Cellulose acetate butyrate,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing