Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carboxylic acid-poly(ethylene glycol)-
b
-poly(D,L lactide)
Linear Formula:
HO(C
3
H
4
O
2
)
m
(C
2
H
4
O)
n
C
2
H
3
O
2
Products
Brand
Sigma-Aldrich (2)
Color
Physical Form
Polymer Composition
Polymer Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
909289
Carboxylic acid-poly(ethylene glycol)-
b
-poly(D,L lactide),
PEG average M
n
5,000, PDLA average M
n
55,000
Expand
View Pricing
909300
Carboxylic acid-poly(ethylene glycol)-
b
-poly(D,L lactide),
PEG average M
n
5,000, PDLA average M
n
16,000
Expand
View Pricing