Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon - vitreous - 3000c
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
GF60986616
Carbon - vitreous - 3000c,
powder, 300 max. part. size (micron), weight 200 g, condition spherical
Expand
View Pricing
GF08213712
Carbon - vitreous - 3000c,
foam, thickness 12.7 mm, size 100 × 100 mm, bulk density 0.05 g/cm
3
Expand
View Pricing
GF10563814
Carbon - vitreous - 3000c,
foam, thickness 10.0 mm, size 60 × 60 mm, bulk density 0.05 g/cm
3
Expand
View Pricing
GF01461563
Carbon - vitreous - 3000c,
powder, max. particle size 300 micron, weight 50 g, condition spherical
Expand
View Pricing
GF26360748
Carbon - vitreous - 3000c,
powder, max. particle size 300 micron, weight 20 g, condition spherical
Expand
View Pricing
GF48958015
Carbon - vitreous - 3000c,
rod, 5.0 mm diameter, length 50 mm, condition glassy carbon
Expand
View Pricing
GF51221090
Carbon - vitreous - 3000c,
foil, thickness 0.5 mm, size 12.5 × 12.5 mm, condition glassy carbon
Expand
View Pricing
GF06936666
Carbon - vitreous - 3000c,
powder, max. particle size 300 micron, weight 100 g, condition spherical
Expand
View Pricing
GF76306901
Carbon - vitreous - 3000c,
powder, 12 max. part. size (micron), weight 20 g, min. particle size 0.4 micron
Expand
View Pricing
GF95409164
Carbon - vitreous - 3000c,
powder, 12 max. part. size (micron), weight 100 g, min. particle size 0.4 micron
Expand
View Pricing
GF38363272
Carbon - vitreous - 3000c,
powder, max. particle size 12 micron, weight 50 g, min. particle size 0.4 micron
Expand
View Pricing