Skip to Content
Merck
Calix[8]arene technical, ≥90% (CH)

Calix[8]arene

Calix[8]arene technical, ≥90% (CH)
Empirical Formula (Hill Notation):
C56H48O8
CAS Number:
Molecular Weight:
848.98

Products

Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing