
Cadmium sulfate hydrate

Synonym(s):
Cadmium sulfate hydrate
Empirical Formula (Hill Notation):
Cd3O12S3 · 8H2O
CAS Number:
Molecular Weight:
769.54
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing