Skip to Content
Merck
Butyrophenone analytical standard

Butyrophenone

Butyrophenone analytical standard
Synonym(s):
1-Benzoylpropane, 1-Phenyl-1-butanone, NSC 8463, Phenyl propyl ketone, Butyrophenone
Linear Formula:
C6H5COCH2CH2CH3
CAS Number:
Molecular Weight:
148.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing