
Butyric acid

Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6