
Buspirone Related Compound K

Synonym(s):
8-Azaspiro[4.5]decane-7,9-dione, 3,3-Tetramethyleneglutarimide
Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2