Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bruker
®
SampleJet NMR tubes with caps
Products
Product Category
tubes (4)
Manufacturer
Diameter (mm)
Length (mm)
OD (mm)
Cap
Color
Frequency (MHz)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
WIMWG10007SJ
Bruker
®
SampleJet NMR tubes with caps,
L 178 mm, cap, O.D. 5.0 mm
Expand
View Pricing
WIMWG10004SJ
Bruker
®
SampleJet NMR tubes with caps,
L 103.5 mm, O.D. 5.0 mm, cap
Expand
View Pricing
WIMWG30004SJ
Bruker
®
SampleJet NMR tubes with caps,
L 103.5 mm, cap, O.D. 3.0 mm
Expand
View Pricing
WIMWG30007SJ
Bruker
®
SampleJet NMR tubes with caps,
L 178 mm, cap, O.D. 3.0 mm
Expand
View Pricing