Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bromothymol blue
Synonym(s):
Bromothymol blue, 3′,3″-Dibromothymolsulfonphthalein
Empirical Formula (Hill Notation):
C
27
H
28
Br
2
O
5
S
CAS Number:
76-59-5
Molecular Weight:
624.38
EC Number:
200-971-2
Products
Product Category
dyes (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Special Grade
ACS reagent (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.03026
Bromothymol blue,
pH indicator, ACS, Reag. Ph Eur
Expand
View Pricing
114413
Bromothymol blue,
ACS reagent, Dye content 95 %
Expand
View Pricing