Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form
Products
Product Category
cylinders (9)
Manufacturer
Volume (ml)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
BR32854
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 500 mL
Expand
View Pricing
BR32862
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 1000 mL
Expand
View Pricing
BR32838
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 100 mL
Expand
View Pricing
BR32848
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 250 mL
Expand
View Pricing
BR32805
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 5 mL
Expand
View Pricing
BR32820
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 25 mL
Expand
View Pricing
BR32808
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 10 mL
Expand
View Pricing
BR32864
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 2000 mL
Expand
View Pricing
BR32828
BRAND
®
USP BLAUBRAND
®
class A measuring cylinder, tall form,
volume 50 mL
Expand
View Pricing