Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
strips of 8 PCR tubes
Products
Product Category
tubes (7)
Manufacturer
Binding Type
Capacity (mL)
Working Volume (ml)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
BR781320
BRAND
®
strips of 8 PCR tubes,
colorless, without caps
Expand
View Pricing
BR781330
BRAND
®
strips of 8 PCR tubes,
colorless, attached caps
Expand
View Pricing
BR781325
BRAND
®
strips of 8 PCR tubes,
polypropylene tube, white, capacity 0.2 mL
Expand
View Pricing
BR781324
BRAND
®
strips of 8 PCR tubes,
blue, without caps
Expand
View Pricing
BR781322
BRAND
®
strips of 8 PCR tubes,
yellow, without caps
Expand
View Pricing
BR781321
BRAND
®
strips of 8 PCR tubes,
rose, without caps
Expand
View Pricing
BR781323
BRAND
®
strips of 8 PCR tubes,
green, without caps
Expand
View Pricing