Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Seal-R-film
™
Sealing Film
Products
Boiling Point (°C)
Manufacturer
Melting Point (°C)
Physical Form
Length (m)
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
BR701623
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W 75 m (50) mm, Single Roll
Expand
View Pricing
BR701621
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W 38 m × 100 mm, Single Roll
Expand
View Pricing
BR701625
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W 75 m × 100 mm, Single Roll
Expand
View Pricing
BR701620
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W (38 m) (100 mm)
Expand
View Pricing
BR701627
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W 15 m × 500 mm, Single Roll
Expand
View Pricing
BR701624
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W 75 m × 100 mm
Expand
View Pricing
BR701622
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W (75 m) (50 mm)
Expand
View Pricing
BR701626
BRAND
®
Seal-R-film
™
Sealing Film,
roll L × W (15 m) (500 mm)
Expand
View Pricing