Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
scoop
Products
Product Category
spatulas/scoops (7)
Manufacturer
Length (mm)
Volume (ml)
Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
BR139135
BRAND
®
scoop,
volume 1000 mL, L 350 mm
Expand
View Pricing
BR139125
BRAND
®
scoop,
volume 250 mL, L 350 mm
Expand
View Pricing
BR139035
BRAND
®
scoop,
volume 500 mL, L 350 mm
Expand
View Pricing
BR139120
BRAND
®
scoop,
volume 100 mL, L 350 mm
Expand
View Pricing
BR139115
BRAND
®
scoop,
volume 50 mL, L 160 mm
Expand
View Pricing
BR139130
BRAND
®
scoop,
volume 500 mL, L 350 mm
Expand
View Pricing
BR139040
BRAND
®
scoop,
volume 1000 mL, L 350 mm
Expand
View Pricing