Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
sample vial
Products
Product Category
vials (5)
Manufacturer
Capacity (mL)
Volume (ml)
Color
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
BR722055
BRAND
®
sample vial,
for Coulter-Counter, volume 20 mL, with lid
Expand
View Pricing
BR115016
BRAND
®
sample vial,
for Technicon-Anal., capacity 2.0 mL
Expand
View Pricing
BR115015
BRAND
®
sample vial,
for Technicon-Anal., capacity 1.5 mL
Expand
View Pricing
BR722060
BRAND
®
sample vial,
for Leuko-Ery Counter, capacity 12 mL, with lid
Expand
View Pricing
BR115017
BRAND
®
sample vial,
for Technicon-Anal., capacity 4.0 mL
Expand
View Pricing