Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
micro tubes without tamper-evident screw cap
Products
Product Category
tubes (5)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
BR780761
BRAND
®
micro tubes without tamper-evident screw cap,
capacity 1.5 mL, self-standing bottom, non-sterile
Expand
View Pricing
BR780760
BRAND
®
micro tubes without tamper-evident screw cap,
capacity 0.5 mL, self-standing bottom, non-sterile
Expand
View Pricing
BR780763
BRAND
®
micro tubes without tamper-evident screw cap,
capacity 2 mL, self-standing bottom, non-sterile
Expand
View Pricing
BR780764
BRAND
®
micro tubes without tamper-evident screw cap,
capacity 2 mL, conical bottom, non-sterile
Expand
View Pricing
BR780762
BRAND
®
micro tubes without tamper-evident screw cap,
capacity 1.5 mL, conical bottom, non-sterile
Expand
View Pricing