Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
microplate BRANDplates
®
, pureGrade
Products
Product Category
plates (32)
Color
Manufacturer
Binding Type
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 32
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
BR781602
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 350 μL, flat bottom, transparent
Expand
View Pricing
BR781620
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 384 wells, size 100 μL, flat bottom, transparent
Expand
View Pricing
BR781608
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 350 μL, flat bottom, black
Expand
View Pricing
BR781660
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 330 μL, round bottom, transparent
Expand
View Pricing
BR781671
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 330 μL, flat bottom, black, with transparent bottom
Expand
View Pricing
BR781607
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 330 μL, round bottom, black
Expand
View Pricing
BR781661
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 360 μL, V-bottom, transparent
Expand
View Pricing
BR781601
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 360 μL, V-bottom, transparent
Expand
View Pricing
BR781621
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 384 wells, size 100 μL, flat bottom, white
Expand
View Pricing
BR781622
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 384 wells, size 100 μL, flat bottom, black
Expand
View Pricing
BR781670
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 330 μL, flat bottom, white, with transparent bottom
Expand
View Pricing
BR781668
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 350 μL, flat bottom, black
Expand
View Pricing
BR781611
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 330 μL, flat bottom, black, with transparent bottom
Expand
View Pricing
BR781605
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 350 μL, flat bottom, white
Expand
View Pricing
BR781600
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 330 μL, round bottom, transparent
Expand
View Pricing
BR781614
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 350 μL, foil bottom, transparent
Expand
View Pricing
BR781665
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 350 μL, flat bottom, white
Expand
View Pricing
BR781604
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 330 μL, round bottom, white
Expand
View Pricing
BR781681
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 384 wells, size 100 μL, flat bottom, white
Expand
View Pricing
BR781615
BRAND
®
microplate BRANDplates
®
, pureGrade,
size 96 wells, size 350 μL, foil bottom, black
Expand
View Pricing
Page 1 of 2
Page 1 of 2