Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
microplate BRANDplates
®
, inertGrade
Products
Product Category
plates (6)
Color
Manufacturer
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR781900
BRAND
®
microplate BRANDplates
®
, inertGrade,
size 96 wells, size 330 μL, round bottom, transparent
Expand
View Pricing
BR781910
BRAND
®
microplate BRANDplates
®
, inertGrade,
size 96 wells, size 330 μL, flat bottom, white, with transparent bottom
Expand
View Pricing
BR781911
BRAND
®
microplate BRANDplates
®
, inertGrade,
size 96 wells, size 330 μL, flat bottom, black, with transparent bottom
Expand
View Pricing
BR781913
BRAND
®
microplate BRANDplates
®
, inertGrade,
size 96 wells, size 330 μL, flat bottom, black, with transparent bottom
Expand
View Pricing
BR781902
BRAND
®
microplate BRANDplates
®
, inertGrade,
size 96 wells, size 350 μL, flat bottom, transparent
Expand
View Pricing
BR781912
BRAND
®
microplate BRANDplates
®
, inertGrade,
size 96 wells, size 330 μL, flat bottom, white, with transparent bottom
Expand
View Pricing