Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale
Products
Product Category
cylinders (16)
Color
Manufacturer
Height (mm)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z327654
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 1,000 mL,
embossed scale: yes
Expand
View Pricing
Z327603
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 250 mL,
embossed scale: yes
Expand
View Pricing
Z327689
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 2,000 mL,
embossed scale: yes
Expand
View Pricing
Z327697
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 2,000 mL, blue graduations
Expand
View Pricing
Z327514
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 10 mL,
embossed scale: yes
Expand
View Pricing
Z327565
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 50 mL, blue graduations
Expand
View Pricing
Z327581
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 100 mL, blue graduations
Expand
View Pricing
Z327670
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 1,000 mL, blue graduations
Expand
View Pricing
Z327530
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 25 mL,
embossed scale: yes
Expand
View Pricing
Z327646
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 500 mL, blue graduations
Expand
View Pricing
Z327549
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 25 mL, blue graduations
Expand
View Pricing
Z327638
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 500 mL,
embossed scale: yes
Expand
View Pricing
Z327573
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 100 mL,
embossed scale: yes
Expand
View Pricing
Z327557
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 50 mL,
embossed scale: yes
Expand
View Pricing
Z327522
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 10 mL, blue graduations
Expand
View Pricing
Z327611
BRAND
®
graduated cylinder, PP, with blue printed scale or embossed scale,
volume 250 mL, blue graduations
Expand
View Pricing