Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
filter tips, racked, TipRack
Products
Product Category
pipette tips (6)
Manufacturer
Volume (µL)
Color
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z740015
BRAND
®
filter tips, racked, TipRack,
volume 0.5-10 μL, BIO-CERT
®
Expand
View Pricing
Z740065
BRAND
®
filter tips, racked, TipRack,
volume 50-1000 μL, BIO-CERT
®
Expand
View Pricing
Z740023
BRAND
®
filter tips, racked, TipRack,
volume 5-100 μL, BIO-CERT
®
Expand
View Pricing
Z740025
BRAND
®
filter tips, racked, TipRack,
volume 5-200 μL, BIO-CERT
®
Expand
View Pricing
Z740017
BRAND
®
filter tips, racked, TipRack,
volume 1-20 μL, BIO-CERT
®
Expand
View Pricing
Z740019
BRAND
®
filter tips, racked, TipRack,
volume 2-20 μL, BIO-CERT
®
Expand
View Pricing