Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
culture tube, rimless
Products
Product Category
tubes (6)
Manufacturer
Capacity (mL)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR114115
BRAND
®
culture tube, rimless,
L × W 16 mm × 100 mm, thickness 0.8 mm
Expand
View Pricing
BR114125
BRAND
®
culture tube, rimless,
L × W 16 mm × 160 mm, thickness 0.8 mm
Expand
View Pricing
BR114130
BRAND
®
culture tube, rimless,
L × W 18 mm × 180 mm, thickness 0.8 mm
Expand
View Pricing
BR114110
BRAND
®
culture tube, rimless,
L × W 12 mm × 100 mm, thickness 0.6 mm
Expand
View Pricing
BR114106
BRAND
®
culture tube, rimless,
L × W 12 mm × 75 mm, thickness 0.6 mm
Expand
View Pricing
BR114105
BRAND
®
culture tube, rimless,
L × W 10 mm × 75 mm, thickness 0.6 mm
Expand
View Pricing