Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
cryogenic tube with screw cap
Products
Product Category
tubes (12)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
BR114832
BRAND
®
cryogenic tube with screw cap,
with external thread self-standing, self-standing, capacity 2 mL
Expand
View Pricing
BR114835
BRAND
®
cryogenic tube with screw cap,
capacity 5 mL, with external thread, self-standing
Expand
View Pricing
BR114833
BRAND
®
cryogenic tube with screw cap,
capacity 3 mL, with external thread, self-standing
Expand
View Pricing
BR114841
BRAND
®
cryogenic tube with screw cap,
capacity 2 mL, Internal thread, self-standing
Expand
View Pricing
BR114840
BRAND
®
cryogenic tube with screw cap,
capacity 1.2 mL, Internal thread, self-standing
Expand
View Pricing
BR114830
BRAND
®
cryogenic tube with screw cap,
capacity 1.2 mL, with external thread, U-bottom bottom (self-standing)
Expand
View Pricing
BR114844
BRAND
®
cryogenic tube with screw cap,
capacity 4 mL, with internal thread self-standing, self-standing
Expand
View Pricing
BR114831
BRAND
®
cryogenic tube with screw cap,
capacity 2 mL, with external thread, round bottom
Expand
View Pricing
BR114834
BRAND
®
cryogenic tube with screw cap,
capacity 4 mL, with external thread, self-standing
Expand
View Pricing
BR114845
BRAND
®
cryogenic tube with screw cap,
capacity 5 mL, with internal thread, round bottom
Expand
View Pricing
BR114842
BRAND
®
cryogenic tube with screw cap,
capacity 2 mL, Internal thread, round bottom
Expand
View Pricing
BR114843
BRAND
®
cryogenic tube with screw cap,
capacity 4 mL, with internal thread, round bottom
Expand
View Pricing