Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
compact automatic BLAUBRAND
®
burette, without support/clamp
Products
Product Category
burettes (3)
Manufacturer
Length (mm)
Volume (ml)
Glassware Class
Color
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BR23821
BRAND
®
compact automatic BLAUBRAND
®
burette, without support/clamp,
volume 50 mL, accuracy: ±0.05 mL, with PE-LD bottle 1000 mL with pedestal, filling tube of PVC
Expand
View Pricing
BR23819
BRAND
®
compact automatic BLAUBRAND
®
burette, without support/clamp,
volume 10 mL, accuracy: ±0.02 mL, with PE-LD bottle 1000 mL with pedestal, filling tube of PVC
Expand
View Pricing
BR23820
BRAND
®
compact automatic BLAUBRAND
®
burette, without support/clamp,
volume 25 mL, accuracy: ±0.03 mL, with PE-LD bottle 1000 mL with pedestal, filling tube of PVC
Expand
View Pricing