Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal
Products
Product Category
flasks (7)
Manufacturer
Color
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
BR36402
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal,
capacity 2 mL, polypropylene stopper, joint: ST/NS 7/16
Expand
View Pricing
BR36404
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal,
capacity 10 mL, polypropylene stopper, joint: ST/NS 10/19
Expand
View Pricing
BR36401
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal,
capacity 1 mL, polypropylene stopper
Expand
View Pricing
BR36408
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal,
capacity 50 mL, polypropylene stopper, joint: ST/NS 12/21
Expand
View Pricing
BR36403
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal,
capacity 5 mL, polypropylene stopper, joint: ST/NS 7/16
Expand
View Pricing
BR36407
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal,
capacity 25 mL, polypropylene stopper, joint: ST/NS 10/19
Expand
View Pricing
BR36406
BRAND
®
BLAUBRAND
®
volumetric flask trapezoidal,
capacity 20 mL, polypropylene stopper, joint: ST/NS 10/19
Expand
View Pricing