Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX)
Products
Product Category
pipettes (29)
Manufacturer
Capacity (mL)
Glassware Class
Package Size
Filter Products
Showing 1-20 of 29
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z325066
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 25 mL, one mark
Expand
View Pricing
Z324930
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 2.5 mL, one mark
Expand
View Pricing
Z324965
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 5 mL, one mark
Expand
View Pricing
Z325023
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 10 mL, one mark
Expand
View Pricing
Z325295
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 1 mL, two marks
Expand
View Pricing
Z325058
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
one mark, capacity 20 mL
Expand
View Pricing
Z325090
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 50 mL, one mark
Expand
View Pricing
Z324914
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 1 mL, one mark
Expand
View Pricing
Z324957
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 4 mL, one mark
Expand
View Pricing
Z324949
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 3 mL, one mark
Expand
View Pricing
Z325333
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 10 mL, two marks
Expand
View Pricing
Z325325
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 5 mL, two marks
Expand
View Pricing
Z325104
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 100 mL, one mark
Expand
View Pricing
Z325317
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 3 mL, two marks
Expand
View Pricing
Z325007
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 8 mL, one mark
Expand
View Pricing
Z324973
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 6 mL, one mark
Expand
View Pricing
Z325015
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
one mark, capacity 9 mL
Expand
View Pricing
Z324906
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 0.5 mL, one mark
Expand
View Pricing
Z324922
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
capacity 2 mL, one mark
Expand
View Pricing
Z324981
BRAND
®
BLAUBRAND
®
bulb pipette, calibrated to deliver (TD, EX),
one mark, capacity 7 mL
Expand
View Pricing
Page 1 of 2
Page 1 of 2