Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brady
®
thermal labeling system (TLS) labels
Products
Product Category
labels (29)
Manufacturer
Color
Package Size
Filter Products
Showing 1-20 of 29
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z663360
Brady
®
thermal labeling system (TLS) labels,
label for 0.2 mL PCR tube, W × L 38.10 mm × 17.28 mm, polyester
Expand
View Pricing
Z663778
Brady
®
thermal labeling system (TLS) labels,
label for 1.0-2.0 mL vial/tube, W × L 41.28 mm × 15.24 mm, polyester
Expand
View Pricing
Z663654
Brady
®
thermal labeling system (TLS) labels,
tamper resistant bottle labels, W × L 76.20 mm × 48.26 mm, white vinyl
Expand
View Pricing
Z663581
Brady
®
thermal labeling system (TLS) labels,
label for 50 mL conical tube, bottle, petri dish or deep well plate, W × L 38.10 mm × 19.05 mm, white Nylon
Expand
View Pricing
Z663603
Brady
®
thermal labeling system (TLS) labels,
tamper resistant bottle labels, W × L 38.10 mm × 25.40 mm, white vinyl
Expand
View Pricing
Z663409
Brady
®
thermal labeling system (TLS) labels,
label for 50 mL conical tube, bottle, petri dish or deep well plate, W × L 25.40 mm × 6.35 mm, white polyester
Expand
View Pricing
Z663905
Brady
®
thermal labeling system (TLS) labels,
vial top label for 1.0 - 2.0 mL vial/tube, writing surface diam. 12.70 mm, yellow Nylon
Expand
View Pricing
Z663883
Brady
®
thermal labeling system (TLS) labels,
vial top label for 0.6 mL vial/tube, writing surface diam. 9.52 mm, yellow Nylon
Expand
View Pricing
Z663484
Brady
®
thermal labeling system (TLS) labels,
label for 50 mL conical tube, bottle, petri dish or deep well plate, W × L 25.40 mm × 12.70 mm, white vinyl
Expand
View Pricing
Z663689
Brady
®
thermal labeling system (TLS) labels,
label for 0.5-0.7 mL centrifuge tube, W × L 45.72 mm × 12.70 mm, polyester
Expand
View Pricing
Z663859
Brady
®
thermal labeling system (TLS) labels,
nameplate / rating label, W × L 48.26 mm × 6.35 mm, polyester
Expand
View Pricing
Z663395
Brady
®
thermal labeling system (TLS) labels,
label for 50 mL conical tube, bottle, petri dish or deep well plate, W × L 25.40 mm × 4.75 mm, white polyester
Expand
View Pricing
Z663646
Brady
®
thermal labeling system (TLS) labels,
label for 50 mL conical tube, bottle, petri dish or deep well plate, W × L 38.10 mm × 25.40 mm, Nylon
Expand
View Pricing
Z663662
Brady
®
thermal labeling system (TLS) labels,
general industrial, W × L 25.40 mm × 15 m, orange vinyl
Expand
View Pricing
Z663670
Brady
®
thermal labeling system (TLS) labels,
general industrial, W × L 25.40 mm × 15 m, yellow vinyl
Expand
View Pricing
Z663700
Brady
®
thermal labeling system (TLS) labels,
label for 50 mL conical tube, bottle, petri dish or deep well plate, W × L 12.70 mm × 12.70 mm, white polyester
Expand
View Pricing
Z663743
Brady
®
thermal labeling system (TLS) labels,
label for 1.0-2.0 mL vial/tube, W × L 66.68 mm × 25.40 mm, polyester
Expand
View Pricing
Z663875
Brady
®
thermal labeling system (TLS) labels,
vial top label for 0.6 mL vial/tube, writing surface diam. 9.52 mm, Nylon
Expand
View Pricing
Z663530
Brady
®
thermal labeling system (TLS) labels,
nameplate / rating label, W × L 76.20 mm × 25.40 mm, white polyester
Expand
View Pricing
Z663611
Brady
®
thermal labeling system (TLS) labels,
nameplate / rating label, W × L 38.10 mm × 25.40 mm, white polyester
Expand
View Pricing
Page 1 of 2
Page 1 of 2