Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brady
®
LABXPERT
™
tapes
Products
Manufacturer
Length (m)
Color
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z664642
Brady
®
LABXPERT
™
tapes,
polyester (continuous), W × L 25.40 mm × 9.1 m
Expand
View Pricing
Z664626
Brady
®
LABXPERT
™
tapes,
polyester (short-tail), W × L 25.40 mm × 44.45 mm , printable L 25.4 mm
Expand
View Pricing
Z664758
Brady
®
LABXPERT
™
tapes,
Nylon (oblong), W × L 31.75 mm × 38.10 mm
Expand
View Pricing