Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BPE-DNPH solid phase extraction (SPE) Cartridge
EC Number:
231-545-4
Products
Brand
Supelco (2)
Agency
Chemical Composition
Volume (ml)
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
54279-U
BPE-DNPH solid phase extraction (SPE) Cartridge,
volume 3 mL, bed B 270 mg (DNPH-coated silica), bed A 130 mg (2-BPE coated silica), pkg of 50 ea
Expand
View Pricing
54278-U
BPE-DNPH solid phase extraction (SPE) Cartridge,
bed B 270 mg (DNPH-coated silica), bed A 130 mg (2-BPE coated silica), volume 3 mL, pkg of 10 ea
Expand
View Pricing