Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boron carbide-hot-pressed
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF84958475
Boron carbide-hot-pressed,
sheet, thickness 1.0 mm, size 25 × 25 mm
Expand
View Pricing
GF89598739
Boron carbide-hot-pressed,
rod, 6.0 mm diameter, length 10 mm
Expand
View Pricing
GF65297111
Boron carbide-hot-pressed,
sheet, thickness 2.0 mm, size 50 × 50 mm
Expand
View Pricing
GF60759647
Boron carbide-hot-pressed,
sheet, thickness 0.5 mm, size 25 × 25 mm
Expand
View Pricing
GF62139876
Boron carbide-hot-pressed,
sheet, thickness 4.0 mm, size 50 × 50 mm
Expand
View Pricing
GF67484155
Boron carbide-hot-pressed,
rod, 6.0 mm diameter, length 100 mm
Expand
View Pricing