Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boric acid
Linear Formula:
H
3
BO
3
CAS Number:
10043-35-3
Molecular Weight:
61.83
Products
Product Category
buffers (6)
acids (2)
inorganic acids (2)
preservatives (2)
salts (1)
Brand
SAFC (3)
Sigma-Aldrich (9)
Supelco (3)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
certified reference material (2)
ACS reagent (1)
Puriss P.A. (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
B0394
Boric acid,
ACS reagent, ≥99.5%
Expand
View Pricing
B0252
Boric acid,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
B6768
Boric acid,
BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Expand
View Pricing
1.00165
Boric acid,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
31146
Boric acid,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Expand
View Pricing
1.00160
Boric acid,
crystalline, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, NF
Expand
View Pricing
B7901
Boric acid,
suitable for electrophoresis, ≥99.5%
Expand
View Pricing
202878
Boric acid,
99.999% trace metals basis
Expand
View Pricing
1.00162
Boric acid,
powder, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, NF
Expand
View Pricing
15663
Boric acid,
BioUltra, for molecular biology, ≥99.5% (T)
Expand
View Pricing
339067
Boric acid,
99.97% trace metals basis
Expand
View Pricing
1.00765
Boric acid,
99.9999 Suprapur
®
Expand
View Pricing
1076341
Boric acid,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
B7660
Boric acid,
BioXtra, ≥99.5%
Expand
View Pricing
1.00164
Boric acid,
cryst., EMPROVE
®
API, Ph. Eur., BP, NF
Expand
View Pricing
1.32264
Boric acid,
powder, Technipur
®
Expand
View Pricing
B2645
Boric acid,
tablet, 1 g boric acid per tablet
Expand
View Pricing
PHR2664
Boric acid,
pharmaceutical secondary standard, certified reference material
Expand
View Pricing
NIST951A
Boric acid,
NIST
®
SRM
®
951a, isotopic standard
Expand
View Pricing
1.32273
Boric acid,
crystals, Technipur
®
Expand
View Pricing